Assay Details
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035087.2 | NP_001030259.2 | 2 - 3 | 228 | 72 | |
GenBank mRNA | BC102368.1 | - | 2 - 3 | 228 | 72 | |
DT819906.1 | - | 2 - 3 | 228 | 72 | ||
DV892281.1 | - | 1 - 2 | 118 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
