Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046074.2 | NP_001039539.1 | 1 - 2 | 403 | 69 | |
GenBank mRNA | BC113343.1 | - | 1 - 2 | 382 | 69 | |
EE252432.1 | - | 1 - 2 | 404 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC113343.1, NM_001046074.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
