Assay Details
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075855.3 | NP_001069323.2 | 3 - 4 | 551 | 129 | |
NR_132766.1 | 3 - 4 | 551 | 129 | |||
GenBank mRNA | BC120438.1 | - | 3 - 4 | 537 | 129 | |
DT720346.1 | - | 3 - 4 | 386 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
