Assay Details
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098013.1 | NP_001091482.1 | 4 - 5 | 696 | 91 | |
GenBank mRNA | BC140517.1 | - | 4 - 5 | 696 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
