Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098040.2 | NP_001091509.2 | 1 - 2 | 146 | 103 | |
GenBank mRNA | BC140495.1 | - | 1 - 2 | 44 | 103 | |
DV927834.1 | - | 1 - 2 | 146 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC140495.1, NM_001098040.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
