Assay Details
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099010.2 | NP_001092480.1 | 2 - 3 | 489 | 87 | |
GenBank mRNA | BC142528.1 | - | 2 - 3 | 490 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
