Assay Details
Gene Symbol: TNFSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101855.2 | NP_001095325.1 | 4 - 5 | 507 | 124 | |
GenBank mRNA | BC149518.1 | - | 4 - 5 | 508 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
