Assay Details
Gene Symbol: DGAT2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101861.1 | NP_001095331.2 | 4 - 5 | 532 | 69 | |
GenBank mRNA | BC149181.1 | - | 4 - 5 | 532 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
