Assay Details
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102038.1 | NP_001095508.1 | 11 - 12 | 1323 | 93 | |
GenBank mRNA | BC148039.1 | - | 11 - 12 | 1323 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
