Assay Details
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102038.1 | NP_001095508.1 | 5 - 6 | 544 | 101 | |
GenBank mRNA | BC148039.1 | - | 5 - 6 | 544 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
