Assay Details
Gene Symbol: CARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103249.1 | NP_001096719.1 | 17 - 18 | 1992 | 93 | |
GenBank mRNA | BC114705.1 | - | 17 - 18 | 1992 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
