Assay Details
Gene Symbol: COP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103256.1 | NP_001096726.1 | 14 - 15 | 1712 | 73 | |
GenBank mRNA | BC133612.1 | - | 14 - 15 | 1712 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
