Assay Details
Gene Symbol: ASPSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104980.1 | NP_001098450.1 | 1 - 2 | 116 | 103 | |
GenBank mRNA | BC142388.1 | - | 1 - 2 | 116 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
