Assay Details
Gene Symbol: HDGFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105361.1 | NP_001098831.1 | 3 - 4 | 352 | 74 | |
GenBank mRNA | BC153286.1 | - | 3 - 4 | 352 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC153286.1, NM_001105361.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
