Assay Details
Gene Symbol: HDGFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105361.1 | NP_001098831.1 | 6 - 7 | 733 | 96 | |
GenBank mRNA | BC153286.1 | - | 6 - 7 | 733 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
