Assay Details
Gene Symbol: CRYBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109791.1 | NP_001103261.1 | 3 - 4 | 2743 | 60 | |
GenBank mRNA | BC149807.1 | - | 3 - 4 | 2743 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
