Assay Details
Gene Symbol: PRMT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099051.1 | NP_001092521.1 | 11 - 12 | 2524 | 67 | |
GenBank mRNA | BC142231.1 | - | 11 - 12 | 2524 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
