Assay Details
Gene Symbol: PRMT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099051.1 | NP_001092521.1 | 2 - 3 | 538 | 110 | |
GenBank mRNA | BC142231.1 | - | 2 - 3 | 538 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
