Assay Details
Gene Symbol: LYPLAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099059.1 | NP_001092529.1 | 3 - 4 | 399 | 130 | |
GenBank mRNA | BC142402.1 | - | 3 - 4 | 399 | 130 | |
BC146087.1 | - | 3 - 4 | 370 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
