Assay Details
Gene Symbol: FBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099063.1 | NP_001092533.1 | 5 - 6 | 1125 | 80 | |
GenBank mRNA | BC142300.1 | - | 5 - 6 | 1125 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
