Assay Details
Gene Symbol: FBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099063.1 | NP_001092533.1 | 6 - 7 | 1315 | 138 | |
GenBank mRNA | BC142300.1 | - | 6 - 7 | 1315 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
