Assay Details
Gene Symbol: MAPK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099080.2 | NP_001092550.1 | 4 - 5 | 2441 | 100 | |
GenBank mRNA | BC142510.1 | - | 4 - 5 | 2442 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
