Assay Details
Gene Symbol: SRD5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099137.1 | NP_001092607.1 | 3 - 4 | 613 | 121 | |
GenBank mRNA | BC142219.1 | - | 3 - 4 | 613 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
