Assay Details
Gene Symbol: EMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099143.2 | NP_001092613.1 | 1 - 2 | 249 | 128 | |
GenBank mRNA | BC134707.1 | - | 1 - 2 | 258 | 128 | |
BC142029.1 | - | 1 - 2 | 250 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
