Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099157.1 | NP_001092627.1 | 1 - 2 | 129 | 125 | |
GenBank mRNA | BC146171.1 | - | 1 - 2 | 129 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC146171.1, NM_001099157.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
