Assay Details
Gene Symbol: PPP1R14D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099169.2 | NP_001092639.1 | 2 - 3 | 572 | 150 | |
GenBank mRNA | BC142353.1 | - | 2 - 3 | 573 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
