Assay Details
Gene Symbol: KRTCAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101209.1 | NP_001094679.1 | 546 | 68 | ||
GenBank mRNA | BC149659.1 | - | 546 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
