Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101211.2 | NP_001094681.1 | 1 - 2 | 124 | 109 | |
GenBank mRNA | BC149246.1 | - | 1 - 2 | 125 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
