Assay Details
Gene Symbol: ZNF131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101218.1 | NP_001094688.1 | 4 - 5 | 576 | 96 | |
GenBank mRNA | BC150130.1 | - | 4 - 5 | 576 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
