Assay Details
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101236.1 | NP_001094706.1 | 1 - 2 | 334 | 136 | |
GenBank mRNA | BC151342.1 | - | 1 - 2 | 334 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
