Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102266.1 | NP_001095736.1 | 5 - 6 | 609 | 91 | |
GenBank mRNA | BC148038.1 | - | 5 - 6 | 609 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
