Assay Details
Gene Symbol: IGFBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102300.2 | NP_001095770.1 | 3 - 4 | 733 | 75 | |
GenBank mRNA | BC148134.1 | - | 3 - 4 | 734 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
