Assay Details
Gene Symbol: EIF4E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102306.1 | NP_001095776.1 | 3 - 4 | 363 | 87 | |
GenBank mRNA | BC148114.1 | - | 3 - 4 | 363 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
