Assay Details
Gene Symbol: HRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102313.1 | NP_001095783.1 | 2 - 3 | 2563 | 70 | |
GenBank mRNA | BC149780.1 | - | 2 - 3 | 2563 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
