Assay Details
Gene Symbol: FYB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105414.1 | NP_001098884.1 | 15 - 16 | 2442 | 113 | |
GenBank mRNA | BC134622.1 | - | 15 - 16 | 2442 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
