Assay Details
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105608.2 | NP_001099078.1 | 1 - 2 | 60 | 131 | |
GenBank mRNA | BC142021.1 | - | 1 - 2 | 61 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142021.1, NM_001105608.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
