Assay Details
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098158.1 | NP_001091627.1 | 3 - 4 | 276 | 89 | |
GenBank mRNA | BC140549.1 | - | 3 - 4 | 276 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
