Assay Details
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099393.1 | NP_001092863.1 | 1 - 2 | 79 | 89 | |
GenBank mRNA | BC146206.1 | - | 1 - 2 | 79 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC146206.1, NM_001099393.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
