Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099399.1 | NP_001092869.1 | 2 - 3 | 347 | 77 | |
GenBank mRNA | BC140678.1 | - | 2 - 3 | 347 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
