Assay Details
Gene Symbol: KHNYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102551.1 | NP_001096021.1 | 3 - 4 | 1481 | 63 | |
GenBank mRNA | BC151400.1 | - | 3 - 4 | 1481 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
