Assay Details
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103348.1 | NP_001096818.1 | 15 - 16 | 1579 | 150 | |
GenBank mRNA | BC142179.1 | - | 15 - 16 | 1579 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
