Assay Details
Gene Symbol: ZHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110095.1 | NP_001103565.1 | 2 - 3 | 168 | 152 | |
GenBank mRNA | BC154375.1 | - | 2 - 3 | 168 | 152 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC154375.1, NM_001110095.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
