Assay Details
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113572.1 | NP_001107044.1 | 5 - 6 | 664 | 130 | |
GenBank mRNA | CO878421.1 | - | 5 - 6 | 555 | 130 | |
DN743221.1 | - | 4 - 5 | 628 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
