Assay Details
Gene Symbol: LOC536190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102374.1 | NP_001095844.1 | 2 - 3 | 477 | 141 |
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC123571.1 | - | 2 - 3 | 477 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
