Assay Details
Gene Symbol: OAS1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178108.2 | NP_835209.1 | 2 - 3 | 501 | 139 | |
GenBank mRNA | AB104656.1 | - | 2 - 3 | 501 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
