Assay Details
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076921.1 | NP_001070389.1 | 1 | 153 | 133 | |
GenBank mRNA | BC123707.1 | - | 1 | 153 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123707.1, NM_001076921.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
