Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 17 - 18 | 2149 | 76 | |
GenBank mRNA | GH316016.1 | - | 3 - 4 | 245 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
