Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 1 - 2 | 146 | 69 | |
GenBank mRNA | DY106259.1 | - | 1 - 2 | 147 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143857.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
