Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 29 - 30 | 3627 | 99 | |
GenBank mRNA | EE354110.1 | - | 4 - 5 | 343 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
