Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 2 - 3 | 251 | 77 | |
GenBank mRNA | DY106259.1 | - | 2 - 3 | 252 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
